Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.08573

Sigma-Aldrich

PPACK II, 2TFA

InSolution, ≥95%, Potent and specific irreversible inhibitor of plasma and glandular kallikreins

Synonym(s):

InSolution PPACK II, 2TFA, D-Phenylalanyl-L-phenylalanyl-L-arginine chloromethyl ketone, 2TFA, H-D-Phe-Phe-Arg-chloromethylketone, 2TFA, PPACK II, Trifluoroacetate Salt, PPACK II, Trifluoroacetate Salt InSolution, D-Phenylalanyl-L-prolyl-L-arginine chloromethyl ketone, 2TFA, H-D-Phe-Phe-Arg-chloromethylketone, 2TFA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H33ClN6O3 · 2C2HF3O2
CAS Number:
Molecular Weight:
729.07
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

storage temp.

−70°C

General description

Potent and specific irreversible inhibitor of plasma and glandular kallikreins. Inhibits the cleavage of atrial naturiuretic peptide (ANP 1-126) by kallikreins. Also inhibits adrenal kininogenase, a kallikrein-like enzyme.

Biochem/physiol Actions

Primary Target
thrombin
Reversible: no
Secondary Target
plasma and glandular kallikreins

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Sequence

H-D-Phe-Phe-Arg-chloromethylketone, 2TFA

Physical form

A 10 mM (5 mg/686 µl) sterile-filtered solution of PPACK II, 2TFA (Cat. No. 520219) in H₂O.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 6 months at -20°C.

Other Notes

Byrne, R.E., and Scanu, A.M. 1989. J. Lipid Res.30, 109.
Scicli, G., et al. 1989. Adv. Exp. Med. Biol.247B, 217.
Gibson, T.R., et al. 1987. Endocrinology120, 764.
Kettner, G., and Shaw, E. 1981. Methods Enzymol.80, 826.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service