Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.08119

Sigma-Aldrich

SB 399885 Hydrochloride

Synonym(s):

SB 399885 Hydrochloride, 5-HT₆ Receptor antagonist, SB 399885 Hydrochloride, SB-399885, SB399885

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21Cl2N3O4S · xHCl
CAS Number:
Molecular Weight:
446.35 (free base basis)
UNSPSC Code:
12352200

assay

≥99% (HPLC)

Quality Level

form

solid

potency

1 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mM
water: 100 mM

storage temp.

2-8°C

InChI

1S/C18H21Cl2N3O4S.ClH/c1-26-17-4-3-13(11-16(17)23-7-5-21-6-8-23)28(24,25)22-15-10-12(19)9-14(20)18(15)27-2;/h3-4,9-11,21-22H,5-8H2,1-2H3;1H

InChI key

RNKCEBCFUSXSQE-UHFFFAOYSA-N

General description

A high affinity 5-HT6 Receptor antagonist (pKi = 9.0). Good solubility (to 100 mM in water or DMSO), oral availability, and penetrant to blood brain barrier. Over 200-fold selective for 5-HT6 receptors vs all receptors, ion channels and enzymes tested in Hirst et al. 2006 publication. Shows nootropic, antidepressant and axiolytic effects comparable to well-described drugs such as diazepam, and therefore has been proposed as novel treatment for schizoprenia and Alzheimer′s disease.

Biochem/physiol Actions

Primary Target
5-HT₆

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wesolowska, A. et al. 2007. Pharmacol Rep.59, 664.
Li, Z. et al. 2007. Brain Res.1134, 70.

Hirst, et al. 2006. Eur. J. Pharm.553, 109.
Routledge, C. et al. 2000. Br. J. Pharmacol.130, 1606.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service