Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.05723

Sigma-Aldrich

SCH 23390 Hydrochloride

Synonym(s):

SCH 23390 Hydrochloride, Dopamine Receptor D1/D5 Antagonist, SCH 23390, (R)-(+)-7-Chloro-8-hydroxy-3-methyl­-1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine Hydrochloride, SCH23390, SCH-23390

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18ClNO · xHCl
CAS Number:
Molecular Weight:
287.78 (free base basis)
MDL number:
UNSPSC Code:
12352200

assay

≥99% (HPLC)

Quality Level

form

solid

potency

0.2 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

water: 100 mM

storage temp.

−20°C

InChI

1S/C17H18ClNO.ClH/c1-19-8-7-13-9-16(18)17(20)10-14(13)15(11-19)12-5-3-2-4-6-12;/h2-6,9-10,15,20H,7-8,11H2,1H3;1H/t15-;/m1./s1

InChI key

OYCAEWMSOPMASE-XFULWGLBSA-N

General description

A highly potent and selective dopamine D1-like receptor antagonist with a Ki of 0.2 and 0.3 nM for the D1 and D5 dopamine receptor subtypes, respectively. Also an agonist at 5-HT1C and 5-HT2C receptors in vitro (Ki values are 6.3 nM and 9.3 nM respectively).

Biochem/physiol Actions

Primary Target
D1 and D5

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Splinter, M. Y. 2007. Ann. Pharmacother.41, 285.
Pfeiffer, R. F. 2002. Drugs Aging.19, 561.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria C Tickerhoof et al.
Psychoneuroendocrinology, 113, 104542-104542 (2019-12-22)
Social interaction with unfamiliar individuals is necessary for species-preserving behaviors such as finding mates and establishing social groups. However, social conflict is a potential negative outcome to interaction with a stranger that can be distressing enough to cause an individual

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service