Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.05300

Sigma-Aldrich

RSK2 Inhibitor, RMM-46

Synonym(s):

RSK2 Inhibitor, RMM-46, ( E)-2-Cyano-N-(1-hydroxy-2-methylpropan-2-yl)-3-(3-(3,4,5-trimethoxyphenyl)-1H-indazol-5-yl)acrylamide, RMM46, RSK Inhibitor IV, (E)-2-Cyano-N-(1-hydroxy-2-methylpropan-2-yl)-3-(3-(3,4,5-trimethoxyphenyl)-1H-indazol-5-yl)acrylamide, RMM46, RSK Inhibitor IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26N4O5
CAS Number:
Molecular Weight:
450.49
UNSPSC Code:
12352200

assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable cyanoacrylamide compound that acts as a potent, reversible, and selective inhibitor of ribosomal S6 kinase 2 (RSK2; IC50 = 12 and 2.5 nM for wild-type and T493M RSK2, respectively). Shown to act by covalently modifying Cys436 at the ATP-binding site. Exhibits good selectivity over NEK2 and PLK1 (IC50 = 530 nM and 2.2 µM) and does not affect the activity of a panel of 26 other protein kinases. Also shown to block phosphorylation of mitogen and stress activated kinases (MSK) 1, 2, and 3 in PMA-stimulated COS7 cells (IC50 ~100 nM) and CREB phosphorylation in PMA- or UV-stimulated HeLa cells (IC50 ~ 300 nM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
RSK2
Reversible: yes
Secondary Target
MSK 1, 2 , 3
Target IC50: 12 and 2.5 nM for wild-type and T493M RSK2, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Miller, R.M., et al. 2013. J. Am. Chem. Soc.135, 5298.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service