Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.04276

Sigma-Aldrich

PKC Activator VII, CGK062

Synonym(s):

PKC Activator VII, CGK062, Protein Kinase C Activator VII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H20O7
CAS Number:
Molecular Weight:
408.40
UNSPSC Code:
12352200

assay

≥98% (HPLC)

Quality Level

form

solid

potency

12.2 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

General description

A cell permeable coumarin-containing compound that is shown to activate PKCα activity and induce cellular PKCα membrane translocation (Effective conc. 12.5 to 50 µM) as well as PKCα-mediated signaling events. Effectively affects the viability of CRT-dependent cancer cells (IC50 = 1.62 to 18.6 µM; 48 h) in vitro and suppresses PC3-derived tumor expansion in mice (50 & 100 mg/kg/day; i.p) in vivo, while exhibiting little cytotoxicity toward WI38 normal fibroblasts.
A cell permeable coumarin-containing compound that is shown to activate PKCα activity toward GST-β-catenin or PKC substrate peptide in cell-free kinase assays (12.5 & 25 µM) and induce cellular PKCα membrane translocation (Effective conc. 12.5 to 50 µM in HEK293, PC3, SNU475, and SW480 cultures) as well as PKCα-mediated β-catenin phosphorylation (Ser33/37/45) and subsequent β-catenin degradation in a F-box Ub ligase complex component β-TrCP- (β-transducin repeat-containing protein) dependent, but GSK-3β-independent, manner. Effectively affects the viability of CRT- (β-catenin response transcription) dependent cancer cells (IC50 = 1.62, 5.71, 9.40, 11.41, and 18.60 µM, respectively, in SW480, HCT15, PC3, SNU475, and DLD-1 cultures; 48 h) in vitro and suppresses PC3-derived tumor expansion in mice (23% and 11% of control size, respectively, with 50 or 100 mg/kg daily i.p. dose for 4 wks) in vivo, while exhibiting little cytotoxicity toward WI38 normal fibroblasts (IC50 = 74.83 µM; 48 h).

Biochem/physiol Actions

Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Gwak, J., et al. 2012. PLoS One.7, e46697.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service