Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

482210

Sigma-Aldrich

Nimesulide Analog, Compound 76

A cell-permeable sulfonanilide that is shown to exhibit an antiproliferative effect against SKBR-3, BT474 and MDA-MB-453, and affects MCF-7 and MDA-MB-231 only at much higher concentrations.

Synonym(s):

Nimesulide Analog, Compound 76

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$188.00

$188.00


Estimated to ship onApril 13, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$188.00

About This Item

Empirical Formula (Hill Notation):
C24H32N2O4S
CAS Number:
Molecular Weight:
444.59
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$188.00


Estimated to ship onApril 13, 2025


Request a Bulk Order

Quality Level

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable sulfonanilide that is shown to exhibit an antiproliferative effect against SKBR-3, BT474 and MDA-MB-453, and affects MCF-7 and MDA-MB-231 only at much higher concentrations.
A cell-permeable sulfonanilide that is shown to exhibit an antiproliferative effect against three HER-2-positive breast cancer cell lines, SKBR-3, BT474 and MDA-MB-453 (IC50= 0.9, 2.2, and 4.0 µM, respectively), and affects two HER-2-negative breast cancer cell lines, MCF-7 and MDA-MB-231 only at much higher concentrations (IC50 = 22.1 and 19.6 µM, respectively). The compound is not expected to be a COX-2 inhibitor and its cellular target(s) and mechanism(s) of growth inhibition is yet to be seen.
A cell-permeable sulfonanilide that is shown to exhibit an antiproliferative effect against three HER-2-positive breast cancer cell lines, SKBR-3, BT474 and MDA-MB-453 (IC50= 0.9, 2.2, and 4.0 µM, respectively), and affects two HER-2-negative breast cancer cell lines, MCF-7 and MDA-MB-231 only at much higher concentrations (IC50 = 22.1 and 19.6 µM, respectively). The compound is not expected to be a COX-2 inhibitor and its cellular target(s) and mechanism(s) of growth inhibition is yet to be seen.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C).

Other Notes

Chen, B., et al. 2009. Biochem. Pharmacol.77, 1787.
Su, B., et al. 2008. J. Comb. Chem.10, 475.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service