Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

436390

Sigma-Aldrich

N-Glycan Acceptor Substrate, Dabsylated Tetrapeptide, Asialo, Agalacto, Biantennary

Useful as a substrate for the non-radioactive assaying by MALDI-mass spectrometry of α1,3- and α1,6-Fuc-transferases, β1,4- and β1,3-Gal-transferases, β1,2-xylosyltransferase, and endo-N-acetylglucosaminidases.

Synonym(s):

GnGn-dabsyl-peptide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<5% β1,3-galactosylated GnGn-dabsyl-peptide

color

burnt orange

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

General description

A dabsylated tetrapeptide with the N-glycan having the asialo-, agalacto-substructure of biantennary N-linked oligosaccharides found in a wide range of glycoproteins. Useful as a substrate for the non-radioactive assaying by MALDI-mass spectrometry of α1,3- and α1,6-Fuc-transferases, β1,4- and β1,3-Gal-transferases, β1,2-xylosyltransferase, and endo-N-acetylglucosaminidases.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze at -20°C. Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Leiter, H., et al. 1999. J. Biol. Chem.274, 21830.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Leiter et al.
The Journal of biological chemistry, 274(31), 21830-21839 (1999-07-27)
Substitution of the asparagine-linked GlcNAc by alpha1,3-linked fucose is a widespread feature of plant as well as of insect glycoproteins, which renders the N-glycan immunogenic. We have purified from mung bean seedlings the GDP-L-Fuc:Asn-linked GlcNAc alpha1,3-fucosyltransferase (core alpha1,3-fucosyltransferase) that is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service