Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

416157

Millipore

Isohelenin, Inula sp. - Calbiochem

Synonym(s):

Isohelenin, Inula sp. - Calbiochem, Isoalantolactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O2
Molecular Weight:
232.32
UNSPSC Code:
12352200

assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL
ethanol: 20 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable sesquiterpene lactone with anti-inflammatory properties. Acts as a highly specific, potent, and irreversible inhibitor of NF-κB activation by preventing IκBα degradation. Does not affect the DNA binding activity of activated NF-κB or inhibit Fyn and Src kinase activities. Also reported to inhibit IL-8 gene expression in cultured human respiratory epithelium and iNOS gene expression in cultured rat aortic smooth muscle cells. Shown to protect against endotoxic shock in rats.
A cell-permeable sesquiterpene lactone with anti-inflammatory properties. Acts as a highly specific, potent, irreversible inhibitor of NF-κB activation that acts by preventing IκBα degradation. Does not affect the DNA binding activity of activated NF-κB, or inhibit p59fyn and Src kinase activities. Inhibits IL-8 gene expression in cultured human respiratory epithelium in vitro, and iNOS gene expression in cultured rat aortic smooth muscle cells. Offers protection against endotoxic shock in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
NF-κB activation
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Sheehan, M., et al. 2002. J. Endotoxin. Res.8, 99.
Mazor, R.L., et al. 2000. Cytokine12, 239.
Wong. H.R., and Menendez, I.Y., 1999. Biochem. Biophys. Res. Commun.262, 375.
Hehner, S.P., et al. 1998. J. Biol. Chem.273, 1288.
Bork, P.M., et al. 1997. FEBS Lett.402, 85.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service