Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

371543

Millipore

Guanosine 5′-O-(2-Thiodiphosphate), Trilithium Salt

Non-hydrolyzable GDP analog that competitively inhibits G-protein activation by GTP and GTP analogs. Stimulates adenylate cyclase activity in sarcolemmal membranes by inactivating Gi proteins.

Synonym(s):

Guanosine 5′-O-(2-Thiodiphosphate), Trilithium Salt, GDP-β-S, 3Li

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N5O10P2S · 3Li
CAS Number:
Molecular Weight:
477.07
MDL number:
UNSPSC Code:
12352208

Quality Level

assay

≥85% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C10H15N5O10P2S.3Li/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(24-9)1-23-26(19,20)25-27(21,22)28;;;/h2-3,5-6,9,16-17H,1H2,(H,19,20)(H2,21,22,28)(H3,11,13,14,18);;;/q;3*+1/p-3/t3-,5-,6-,9-;;;/m1.../s1

InChI key

LMCWQGPJYZRMKU-CYCLDIHTSA-K

General description

A nonhydrolyzable GDP analog that competitively inhibits G-protein activation by GTP and GTP analogs.
Non-hydrolyzable GDP analog that competitively inhibits G-protein activation by GTP and GTP analogs. Stimulates adenylate cyclase activity in sarcolemmal membranes by inactivating Gi proteins.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions lose 5% of their activity after 6 months at -20°C.

Other Notes

Piacentini, L., et al. 1996. Biochim. Biophys. Acta1282, 11.
Eberhard, D.A. and Holz, R.W., et al. 1991. Biochem J.279, 447.
Silk, S.T., et al. 1989. J. Biol. Chem.264, 21466.
Burch, R.M., et al. 1987. Proc. Natl. Acad. Sci. USA84, 6374.
Gilman, A.G. 1987. Annu. Rev. Biochem.56, 615.
Eckstein, F., et al. 1979. J. Biol. Chem.254, 9829.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service