Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

361541

Sigma-Aldrich

GSK-3β Inhibitor II - CAS 478482-75-6 - Calbiochem

The GSK-3β Inhibitor II, also referenced under CAS 478482-75-6, controls the biological activity of GSK-3β. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

GSK-3β Inhibitor II - CAS 478482-75-6 - Calbiochem, Glycogen Synthase Kinase-3β Inhibitor II, 2-Thio(3-iodobenzyl)-5-(1-pyridyl)-[1,3,4]-oxadiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10IN3OS
CAS Number:
Molecular Weight:
395.22
UNSPSC Code:
12352200

Quality Level

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A 2-thio-[1,3,4]-oxadiazole-pyridyl derivative that acts as a potent inhibitor of glycogen synthase kinase-3β (IC50 = 390 nM).
A 2-thio-[1,3,4]-oxadiazole-pyridyl derivative that acts as a potent inhibitor of glycogen synthase kinase-3b (IC50 = 390 nM).



Biochem/physiol Actions

Cell permeable: no
Primary Target
Gsk-3β
Product does not compete with ATP.
Reversible: no
Target IC50: 390 nM against glycogen synthase kinase-3β

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Naerum, L., et al. 2002. Bioorg. Med. Chem. Lett.12, 1525.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lars Naerum et al.
Bioorganic & medicinal chemistry letters, 12(11), 1525-1528 (2002-05-29)
Using a virtual screening strategy based on a methodology derived from the CATS molecular descriptor, a novel compound class with inhibitory activity against the GSK-3 enzyme was identified through scaffold hopping. These compounds were readily synthesized, either by solid-phase or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service