Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

343022

Sigma-Aldrich

Flt-3 Inhibitor III

The Flt-3 Inhibitor III, also referenced under CAS 852045-46-6, controls the biological activity of Flt-3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Flt-3 Inhibitor III, (5-Phenyl-thiazol-2-yl)-(4-(2-pyrrolidin-1-yl-ethoxy)-phenyl)-amine, 5-Phenyl-N-(4-(2-(1-pyrrolidinyl)ethoxy)phenyl)-2-thiazolamine, 5-Phenyl-N-(4-(2-(1-pyrrolidinyl)ethoxy)phenyl)-2-thiazolamine, (5-Phenyl-thiazol-2-yl)-(4-(2-pyrrolidin-1-yl-ethoxy)-phenyl)-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23N3OS
CAS Number:
Molecular Weight:
365.49
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable 5-phenyl-2-thiazolamine compound that acts as a potent, ATP binding site-targeting inhibitor of Flt3 (IC50 = 50 nM). It inhibits c-Kit, KDR, c-Abl, Cdk1, c-Src, and Tie-2 only at much higher concentrations (IC50 = 0.26, 0.91, 1.2, 2.1, 2.8, and 8.0 µM, respectively) and exhibits little activity against a panel of 12 other commonly studied kinases even at concentrations as high as 10 µM. Blocks FLT3-dependent cell proliferation in a dose-dependent manner (IC50 = 52, 240, and 760 nM in MV4:11-ITD, BaF3-ITD, and BaF3-D835/Y cells).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Flt-3
Product competes with ATP.
Reversible: no
Target IC50: 50 nM against Flt3

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Furet, P., et al. 2006. J. Med. Chem.49, 4451.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service