Skip to Content
MilliporeSigma
All Photos(1)

Documents

324881

Sigma-Aldrich

PERK, GST-Fusion, Human, Recombinant, E. coli

Synonym(s):

EIF2AK3 PKR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

assay

≥90% (SDS-PAGE)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Note: the calculated MW is 88,000, but the protein runs at ~115,000 on SDS-PAGE.
Recombinant, human PERK consisting of amino acids 563-1115 fused at the N-terminus to GST and expressed in E. coli. PERK phosphorylates the α-subunit of eukaryotic translation initiation factor 2 (EIF2α), which leads to its inactivation and reduces translation initiation. The active kinase is useful for the study of PERK regulation and for inhibitor screening.
Recombinant, human PERK consisting of amino acids 563-1115 fused at the N-terminus to GST and expressed in E. coli. PERK phosphorylates the α-subunit of eukaryotic translation initiation factor 2 (EIF2α), which leads to its inactivation and reduces translation initiation. The active kinase is useful for the study of PERK regulation and for inhibitor screening. Note: the calculated MW is 88,000, but the protein runs at ~115,000 on SDS-PAGE.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

Kinase activity is measured as the molar amount of phosphate incorporated into SMAD3 at 30°C using a final concentration of 50 µM ATP.

Physical form

In 50 mM Tris-HCl, 50 mM NaCl, 250 µM DTT, 100 µM EGTA, 100 µM EDTA, 100 µM PMSF, 25% glycerol, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Baltzis, D., et al. 2007. J Biol Chem.43, 31675.
Koumenis, C., et al. 2002. Mol Cell Biol.21, 7405.


Specific Activity: Please refer to the lot-specific Certificate of Analysis

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service