Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

324380

Millipore

Doxorubicin, Hydrochloride - CAS 25316-40-9 - Calbiochem

Doxorubicin HCl, CAS 25316-40-9, is an antitumor antibiotic that inhibits topoisomerase II (IC₅₀ = 100 nM).

Synonym(s):

Doxorubicin, Hydrochloride - CAS 25316-40-9 - Calbiochem

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29NO11 · xHCl
CAS Number:
Molecular Weight:
543.52 (free base basis)
MDL number:
UNSPSC Code:
12352200

Quality Level

assay

≥98% (HPLC)

form

powder

potency

100 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

orange to red

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C27H29NO11.ClH/c1-10-22(31)13(28)6-17(38-10)39-15-8-27(36,16(30)9-29)7-12-19(15)26(35)21-20(24(12)33)23(32)11-4-3-5-14(37-2)18(11)25(21)34;/h3-5,10,13,15,17,22,29,31,33,35-36H,6-9,28H2,1-2H3;1H/t10?,13?,15-,17?,22?,27-;/m0./s1

InChI key

MWWSFMDVAYGXBV-FGBJBKNOSA-N

General description

An anti-tumor antibiotic and a highly effective myotoxin that inhibits topoisomerase II (IC50 = 100 nM). Binds to nucleic acids, presumably by specific intercalation into the DNA double helix, inhibiting nucleic acid synthesis. Cytotoxicity seems to be due to its ability to intercalate with DNA, interact with plasma membranes, and take part in oxidation-reduction reactions. Induces apoptosis in rhabdomyosarcoma cell lines.
Antitumor antibiotic and a highly effective myotoxin that inhibits topoisomerase II (IC50 = 100 nM). Binds to nucleic acids, presumably by specific intercalation into the DNA double helix, thereby inhibiting nucleic acid synthesis. Induces apoptosis in rhabdomyosarcoma cell lines. Also available as a 10 mM solution in H2O (Cat. No. 504042).
Doxorubicin HCl, CAS 25316-40-9, is an antitumor antibiotic that inhibits topoisomerase II (IC₅₀ = 100 nM).

Biochem/physiol Actions

Primary Target
topoisomerase 2
Product does not compete with ATP.

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 1 month at 4°C. Under acidic conditions doxorubicin is converted to a water-insoluble adriamycinone and a water soluble reducing sugar, daunosamine.

Other Notes

A′Hern, R.P., and Gore, M.E. 1995. J. Clin. Oncol. 13, 726.
Nooter, K., et al. 1995. Br. J. Cancer 71, 556.
Noviello, E., et al. 1994. Mutat. Res.311, 21.
Anderson, R.D., et al. 1993. Mutat. Res.294, 215.
Hershko, C., et al. 1993. J. Lab. Clin. Med.122, 245.
Jongmans, W., et al. 1993. Mutat. Res.294, 207.
O′Shaughnessy, J.A., and Cowan, K.H. 1993. J. Am. Med. Assoc.270, 2089.
Theyer, G., et al. 1993. J. Urol.150, 1544.
Tritton, T.R., and Yee, G. 1982. Science217, 248.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Muta. 1B - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy J Humpton et al.
Genes & development, 35(1-2), 117-132 (2020-12-19)
The p53 tumor suppressor protein is a potent activator of proliferative arrest and cell death. In normal cells, this pathway is restrained by p53 protein degradation mediated by the E3-ubiquitin ligase activity of MDM2. Oncogenic stress releases p53 from MDM2
Pietri Puustinen et al.
Autophagy, 16(10), 1871-1888 (2020-01-28)
Macroautophagy/autophagy is a central component of the cytoprotective cellular stress response. To enlighten stress-induced autophagy signaling, we screened a human kinome siRNA library for regulators of autophagic flux in MCF7 human breast carcinoma cells and identified the catalytic subunit of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service