Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

228230

Sigma-Aldrich

Cholesterol Oxidase, Nocardia sp.

Cholesterol Oxidase, Nocardia sp. CAS 9028-76-6, is a native cholesterol oxidase that catalyzes the oxidation of cholesterol to cholest-4-ene-3-one.

Synonym(s):

Cholesterol Oxidase, Nocardia sp.

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202

Quality Level

form

solid

specific activity

≥10 units/mg dry wt.
>11 U/mg

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

pale brown

solubility

PBS: 1 mg/mL

foreign activity

catalase and glucose oxidase ≤0.1%

shipped in

ambient

storage temp.

−20°C

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service