Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

200496

Sigma-Aldrich

Betacellulin, Human, Recombinant

Recombinant, human betacellulin consisting of amino acids 32-111 and expressed in E. coli. A heparin-binding protein that is able to bind to the EGF receptor.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

Pricing and availability is not currently available.

Quality Level

assay

≥97% (SDS-PAGE)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<1.0 EU/μg protein Endotoxins

shipped in

ambient

storage temp.

−20°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB4300039SAB4300133SAB4300032
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

General description

Recombinant, human betacellulin (BTC) consisting of amino acids 32-111 and expressed in E. coli. BTC is a heparin-binding protein, that is able to bind to the EGF receptor. It is also a potent mitogen for Balb/c 3T3 fibroblasts, retinal pigment epithelial cells and vascular smooth muscle cells.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Reconstitute to 100 µg/ml in sterile PBS containing at least 0.1% human or bovine serum albumin.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service