Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

189728

Sigma-Aldrich

Avidin, Peroxidase Conjugate

This Avidin, Peroxidase Conjugate is validated for use in EIA Immunohistochemistry.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133

Quality Level

form

liquid

contains

≤0.1% thimerosal as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

shipped in

wet ice

storage temp.

2-8°C

General description

Chicken egg white avidin conjugated to horseradish peroxidase. Specific for biotin, biotinylated proteins, or biotinylated antibodies.

Application

EIA (1:1000)

Immunohistochemistry (1:500)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 10 mM PBS, 500 µM EDTA, 10% BSA, 10% mannose, 2% sucrose, pH 8.0.

Other Notes

Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

To comply with ban of sale of mercury-added products required by The Interstate Mercury Education and Reduction Clearinghouse (IMERC), this product is prohibited to be sold in the following US states: Rhode Island and Connecticut.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service