Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

17-655

Sigma-Aldrich

ChIPAb+ Nanog Antibody - ChIP Validated Antibody and Primer Set

from mouse

Synonym(s):

homeobox protein NANOG, homeobox transcription factor Nanog, hNanog

Sign Into View Organizational & Contract Pricing

Select a Size

25 ASSAYS
$580.00

$580.00


Estimated to ship onApril 19, 2025


Request a Bulk Order

Select a Size

Change View
25 ASSAYS
$580.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.75

$580.00


Estimated to ship onApril 19, 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

monoclonal

species reactivity

human

manufacturer/tradename

ChIPAb+
Upstate®

technique(s)

ChIP: suitable
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

All ChIPAb+ antibodies are individually validated for chromatin precipitation, every lot, every time. Each ChIPAb+ antibody set includes control primers (tested every lot by qPCR) to biologically validate your IP results in a locus-specific context. The qPCR protocol and primer sequences are provided, allowing researchers to validate ChIP protocols when using our antibody in their chromatin context. Each set also includes a negative control antibody to ensure specificity of the ChIP reaction.
The ChIPAb+ Nanog set includes a Nanog antibody, a Normal Mouse IgG, and positive control primers which amplify a 81 bp of human miR-302 cluster promoter region. The Nanog antibody and negative control IgG are supplied in a scalable "per ChIP" reaction size and can be used to functionally validate the precipitation of Nanog associated chromatin

Application

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from Ntera-2 cells fixed for 15 min (1e5 cell equivalents per IP) were subjected to chromatin immunoprecipitation using 3 µg of either Normal Mouse IgG (Part No. CS200621), or 3 µg Anti-Nanog (Part No. CS207343) and the Magna ChIP HiSens (Cat. # 17-10460). Successful immunoprecipitation of Nanog associated DNA fragments was verified by qPCR using ChIP Primers, miR-302 (Part No. CS207380) as a positive locus, and Nanog minus 10 kb primers as a negative locus. Data are presented as percent input of each IP sample relative to input chromatin for each amplicon and ChIP sample as indicated.
Please refer to the Magna ChIP HiSens (Cat. # 17-10460) or EZ-MagnaChIP HiSens(Cat. # 17-10461) protocol for experimental details.

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from Ntera-2 cells fixed for 15 min (1e4, 1e5 and 1e6 cell equivalents per IP) were subjected to chromatin immunoprecipitation using 3 µg of either Normal Mouse IgG (Part No. CS200621), or 3 µg Anti-Nanog (Part No. CS207343) and the Magna ChIP HiSens Kit (Cat. # 17-10460). Successful immunoprecipitation of Nanog associated DNA fragments was verified by qPCR using ChIP Primers, miR-302 (Part No. CS207380.
Please refer to the MagnaChIP HiSens (Cat. # 17-10460) or EZ-MagnaChIP HiSens (Cat. # 17-10461) protocol for experimental details.

Western Blotting Analysis:
0.5 µg/mL of this antibody detected NANOG on 10 µg of H9 human embryonic stem cell lysate.

Immunocytochemistry Analysis:
A 1:500 dilution from a representative lot detected NANOG in H9 human embryonic stem cells.
Research Category
Epigenetics & Nuclear Function

Nuclear Function
Use ChIPAb+ Nanog ChIP Validated mouse monoclonal antibody for the detection of Nanog.

Packaging

25 assays per set. Recommended use: 3.0 μg of antibody per chromatin immunoprecipitation (dependent upon biological context).

Quality

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from Ntera-2 cells fixed for 15 min (1e5 cell equivalents per IP) were subjected to chromatin immunoprecipitation using 3 µg of either Normal Mouse IgG (Part No. CS200621), or 3 µg Anti-Nanog (Part No. CS207343) and the Magna ChIP® HiSens Kit (Cat. # 17-10460). Successful immunoprecipitation of Nanog associated DNA fragments was verified by qPCR using ChIP Primers, miR-302 (Part No. CS207380).
Please refer to the MagnaChIP HiSens (Cat. # 17-10460) or EZ-MagnaChIP HiSens (Cat. # 17-10461) protocol for experimental details.

Physical form

Anti-Nanog (Mouse Monoclonal), Part No. CS207343. One vial containing 110 µL (0.7 mg/mL) purified mouse monoclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4),150mM NaCl, and 0.05% sodium azide, before the addition of 30% glycerol. Store at -20°C.
Normal Mouse IgG, Part No. CS200621. One vial containing 125 μg of purified mouse IgG in 125 μL storage buffer containing 0.1% sodium azide. Store at -20°C.
ChIP Primers, miR-302. Part No. CS207380. One vial containing 75 μL of 5 μM of each primer specific for human miR-302 cluster promoter region (chr4:113570181-113570261, hg19 build). Store at -20°C.
FOR: CCC GTG GAA GCA ATC TAT TTA TTT
REV: TGT GTT TCT ATC TGG AGG AAC TCT GT
Format: Purified

Analysis Note

Control
Includes normal mouse IgG and primers specific for human miR-302 cluster promoter region.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MAGNA CHIP is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service