17-10045
ChIPAb+ Acetyl-Histone H4 (Lys5) - ChIP Validated Antibody and Primer Set, rabbit monoclonal
culture supernatant, from rabbit
Synonym(s):
H4K5Ac, Histone H4 (acetyl K5), H4 histone family, member A, histone 1, H4a, histone cluster 1, H4a
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
biological source
rabbit
Quality Level
antibody form
culture supernatant
clone
monoclonal
species reactivity
human
manufacturer/tradename
ChIPAb+
Upstate®
technique(s)
ChIP: suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
dry ice
General description
All ChIPAb+ antibodies are individually validated for chromatin precipitation, every lot, every time. Each ChIPAb+ antibody set includes control primers (tested every lot by qPCR) to biologically validate your IP results in a locus-specific context. The qPCR protocol and primer sequences are provided, allowing researchers to validate ChIP protocols when using our antibody in their chromatin context. Each set also includes a negative control antibody to ensure specificity of the ChIP reaction.
The ChIPAb+ Acetyl-Histone H4 (Lys5) set includes the Acetyl-Histone H4 (Lys5) antibody, a negative control supernatant, and qPCR primers which amplify a 166 bp region of human GAPDH. The Acetyl-Histone H4 (Lys5) and negative controls are supplied in a scalable "per ChIP" reaction size and can be used to functionally validate the precipitation of Acetyl-Histone H4 (Lys5)-associated chromatin.
The ChIPAb+ Acetyl-Histone H4 (Lys5) set includes the Acetyl-Histone H4 (Lys5) antibody, a negative control supernatant, and qPCR primers which amplify a 166 bp region of human GAPDH. The Acetyl-Histone H4 (Lys5) and negative controls are supplied in a scalable "per ChIP" reaction size and can be used to functionally validate the precipitation of Acetyl-Histone H4 (Lys5)-associated chromatin.
Histone H4 is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H4 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. Acetylation of histone H4 occurs at several different lysine positions in the histone tail and is performed by a family of enzymes known as Histone Acetyl Transferases (HATs).
Specificity
Recognizes Histone H4 acetylated on Lys5.
Wide range of cross-reactivity expected based on sequence homology.
Immunogen
Epitope: Acetylated Lys5
Peptide corresponding to Histone H4 containing the sequence [GRG-AcK-GGK] on which Lys5 is acetylated.
Application
Chromatin Immunoprecipitation:
Sonicated chromatin prepared from untreated HeLa cells (1 X 106 cell equivalents per IP) were subjected to chromatin immunoprecipitation using 2 µL of either a negative control supernatant , or 2 µL of Anti-Acetyl-Histone H4 (Lys5) and the Magna ChIP® A Kit (Cat. # 17-610).
Successful immunoprecipitation of acetyl-Histone H4 (Lys5) associated DNA fragments was verified by qPCR using control Primers flanking the human GAPDH promoter, or primers amplifying the promoter of human β-globin, which is transcriptionally inactive in HeLa cells (Please see figures).
Data is presented as percent input of each IP sample relative to input chromatin for each amplicon and ChIP sample as indicated.
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.
Western Blot Analysis:
Representative lot data.
Lysates from HeLa cells untreated or sodium butyrate treated (Lanes 1 and 2 respectively) were probed with anti-acetyl-Histone H4 (Lys5) (1:1,000 dilution).
Proteins were visualized using goat anti-rabbit. (Please see figures).
Western Blot Analysis/Peptide Inhibition: Representative lot data.
Sodium butyrate treated HeLa acid extract was resolved by electrophoresis, transferred to PVDF membrane and probed with Anti-Acetyl-Histone H4 (Lys5) (1:1000 dilution).
To demonstrate specificity, serum was preincubated with modified Histone H4 peptides:
Lane 1: No peptide
Lane 2: Acetyl-Histone H4 (K5) (Cat. # 12-343)
Lane 3: Acetyl-Histone H4 (K8) (Cat. # 12-344)
Lane 4: Acetyl-Histone H4 (K12) (Cat. # 12-345)
Lane 5: Acetyl-Histone H4 (K16) (Cat. # 12-346)
Lane 6: Acetyl-Histone H4 (K5, 8, 12, 16) (Cat. # 12-353)
Lane 7: Histone H4 (Cat. # 12-347)
Proteins were visualized using a donkey anti-rabbit IgG conjugated to HRP and visualized using a chemiluminescence detection system. (Please see figures).
Sonicated chromatin prepared from untreated HeLa cells (1 X 106 cell equivalents per IP) were subjected to chromatin immunoprecipitation using 2 µL of either a negative control supernatant , or 2 µL of Anti-Acetyl-Histone H4 (Lys5) and the Magna ChIP® A Kit (Cat. # 17-610).
Successful immunoprecipitation of acetyl-Histone H4 (Lys5) associated DNA fragments was verified by qPCR using control Primers flanking the human GAPDH promoter, or primers amplifying the promoter of human β-globin, which is transcriptionally inactive in HeLa cells (Please see figures).
Data is presented as percent input of each IP sample relative to input chromatin for each amplicon and ChIP sample as indicated.
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.
Western Blot Analysis:
Representative lot data.
Lysates from HeLa cells untreated or sodium butyrate treated (Lanes 1 and 2 respectively) were probed with anti-acetyl-Histone H4 (Lys5) (1:1,000 dilution).
Proteins were visualized using goat anti-rabbit. (Please see figures).
Western Blot Analysis/Peptide Inhibition: Representative lot data.
Sodium butyrate treated HeLa acid extract was resolved by electrophoresis, transferred to PVDF membrane and probed with Anti-Acetyl-Histone H4 (Lys5) (1:1000 dilution).
To demonstrate specificity, serum was preincubated with modified Histone H4 peptides:
Lane 1: No peptide
Lane 2: Acetyl-Histone H4 (K5) (Cat. # 12-343)
Lane 3: Acetyl-Histone H4 (K8) (Cat. # 12-344)
Lane 4: Acetyl-Histone H4 (K12) (Cat. # 12-345)
Lane 5: Acetyl-Histone H4 (K16) (Cat. # 12-346)
Lane 6: Acetyl-Histone H4 (K5, 8, 12, 16) (Cat. # 12-353)
Lane 7: Histone H4 (Cat. # 12-347)
Proteins were visualized using a donkey anti-rabbit IgG conjugated to HRP and visualized using a chemiluminescence detection system. (Please see figures).
Research Category
Epigenetics & Nuclear Function
Epigenetics & Nuclear Function
Research Sub Category
Histones
Histones
This ChIPAb+ Acetyl-Histone H4 (Lys5) -ChIP Validated Antibody & Primer Set conveniently includes the antibody & the specific control PCR primers.
Packaging
25 assays per set. Recommended use: ~2 μL of antibody per chromatin immunoprecipitation (dependent upon biological context).
Quality
Chromatin Immunoprecipitation:
Sonicated chromatin prepared from untreated HeLa cells (1 X 106 cell equivalents per IP) was subjected to chromatin immunoprecipitation using 2 µL of either negative control supernatant or 2 µL of Anti-Acetyl-Histone H4 (Lys5) and the Magna ChIP® A Kit (Cat. # 17-610).
Successful immunoprecipitation of acetyl-Histone H4 (Lys5) associated DNA fragments was verified by qPCR using control ChIP Primers flanking the human GAPDH promoter (Please see figures).
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.
Sonicated chromatin prepared from untreated HeLa cells (1 X 106 cell equivalents per IP) was subjected to chromatin immunoprecipitation using 2 µL of either negative control supernatant or 2 µL of Anti-Acetyl-Histone H4 (Lys5) and the Magna ChIP® A Kit (Cat. # 17-610).
Successful immunoprecipitation of acetyl-Histone H4 (Lys5) associated DNA fragments was verified by qPCR using control ChIP Primers flanking the human GAPDH promoter (Please see figures).
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.
Target description
~11 kDa
Physical form
Anti-Acetyl-Histone H4 (Lys5) (rabbit monoclonal IgG). One vial containing 50 μL of rabbit monoclonal IgG cell culture supernatant in 0.1% sodium azide. Store at -20°C.
Negative Control Supernatant (rabbit). One vial containing 50 μL of cultured supernatant in 0.05% sodium azide. Store at -20°C.
Control Primers. One vial containing 75 μL of 5 μM each primer specific for the human GAPDH promoter. Store at -20°C.
FOR: TAC TAG CGG TTT TAC GGG CG
REV: TCG AAC AGG AGG AGC AGA GAG CGA
Negative Control Supernatant (rabbit). One vial containing 50 μL of cultured supernatant in 0.05% sodium azide. Store at -20°C.
Control Primers. One vial containing 75 μL of 5 μM each primer specific for the human GAPDH promoter. Store at -20°C.
FOR: TAC TAG CGG TTT TAC GGG CG
REV: TCG AAC AGG AGG AGC AGA GAG CGA
Storage and Stability
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Analysis Note
Control
Includes negative control supernatant and primers specific for human GAPDH.
Includes negative control supernatant and primers specific for human GAPDH.
Legal Information
MAGNA CHIP is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service