Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-699-M

Sigma-Aldrich

PAK5 Protein, active, 10 µg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

mol wt

Mw 37.4 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

N-terminal 6His-tagged, recombinant, human PAK5, amino acids 425-end. PAK5 is also known as PAK7.
expressed by baculovirus in Sf21 cells

Application

Active, N-terminal 6His-tagged, recombinant, human PAK5, amino acids 425-end. PAK5 is also known as PAK7. For use in Kinase Assays.
Research Category
Metabolism
Research Sub Category
Obesity|Metabolic Disorders

Biochem/physiol Actions

  • Protein Target: PAK5
  • Target Sub-Family:STE

Quality

routinely evaluated by phosphorylation of PAKtide

Physical form

Format: Ni2+/NTA-agarose

Storage and Stability

6 months at -70°C from date of shipment

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service