Skip to Content
MilliporeSigma
All Photos(1)

Documents

14-626-M

Sigma-Aldrich

EGFR (L858R) Protein, active, 10 µg

Active, N-terminal GST-tagged, recombinant human EGF Receptor, amino acids 696-end containing the mutation L858R, for use in Enzyme Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 85.8 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

UniProt accession no.

General description

N-terminal GST-tagged, recombinant human EGF Receptor, amino acids 696-end containing the mutation L858R

Quality

routinely evaluated by phosphorylation of 0.1mg/ml Poly (Glu4-Tyr)

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas J Lynch et al.
The New England journal of medicine, 350(21), 2129-2139 (2004-05-01)
Most patients with non-small-cell lung cancer have no response to the tyrosine kinase inhibitor gefitinib, which targets the epidermal growth factor receptor (EGFR). However, about 10 percent of patients have a rapid and often dramatic clinical response. The molecular mechanisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service