Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

144220

Floor Plate

for use with RCS® Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409

material

metal support

agency

non-compendial

description

stability

form

pieces

packaging

pkg of 1 kit

application(s)

air monitoring
food and beverages

compatibility

for use with RCS® Standard

shipped in

ambient

storage temp.

room temp

General description

940020 (RCS Standard)
For using the RCS Standard in an upright position.

Legal Information

RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service