Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.00639

Millipore

Acrylamide-bis ready-to-use solution 30% (37.5:1)

for electrophoresis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105319
This product is sold exclusively by Millipore via offline channels. Contact Customer Service

Quality Level

form

liquid

density

1.03 g/cm3

storage temp.

2-8°C

Analysis Note

Appearance of solution (colour): almost colourless
Appearance of solution (clearness): almost clear
Assay of N,N′-methylene-bisacrylamide (HPLC): 2.4 - 2.8 %
Free acid (as acrylic acid): ≤ 0.002 %
Spec. Absorptivity A 1%/1cm (290 nm; 1 %; 1 cm; water): ≤ 0.15

pictograms

Exclamation markHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

target_organs

Peripheral nervous system

Storage Class

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service