Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03-0182-00

Millipore

SMC® Human IL-12p70 High Sensitivity Immunoassay Kit

Synonym(s):

IL-12p70 Single Molecule Counting Kit, IL-12p70 ultra-sensitive immunoassay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84
Pricing and availability is not currently available.

species reactivity

canine, rat, mouse, feline, monkey

manufacturer/tradename

SMC®

assay range

sensitivity: 0.1 pg/mL
(Lower Limit of Quantification)

inter-assay cv: <20%
intra-assay cv: <15%
(Inter-assay n=5; Intra-assay n=20)

technique(s)

single molecule counting: suitable

storage temp.

2-8°C

Specificity

Species Reactivity:
Although IL-12p70 was detectable in rat, mouse, monkey, canine, and feline plasma, the assay has not been validated for these species.

Cross Reactivity:
Cross-reactivity to interferons of other types was tested using SMC® Human IL-12p70 High Sensitivity Immunoasay Kit with the following results:
  • rhIL-12p40 was found to be < 2.0% cross-reactive
  • rhIL-23 was found to be < 1.5% cross-reactive
  • rhILp35 was not found to be cross-reactive


Accuracy
  • Minimum of 5 serum samples spiked with average recovery of 103% with a range of 100-106%
  • Minimum of 5 plasma samples spiked with average recovery of 97% with a range of 84-102%

Application

The sensitivity of the Single Molecule Counting (SMC®) technology gives the SMC® Human IL-12p70 High Sensitivity Immunoassay Kit unparalleled performance in the quantification of IL-12p70 in human serum and plasma.

This new streamline kit has been improved with the following features over the legacy IL-12 kit, #03-0057-00:
  • Improved options for sample prep and incubation
  • Lyophilization of the standard provides the kit at one storage condition
  • Removal of a plate transfer step
  • Increased reagent volumes to accommodate automation
  • Validated to run on both SMCxPRO® and Erenna
  • Matched to WHO standard


Research Category:
Inflammation & Immunology

Research Sub-Category:
  • Inflammatory Disease
  • Immune Response
  • Autoimmune disease

Packaging

96 well assay

Components

  • Assay Buffer
  • IL-12p70 Coated Beads
  • Standard Diluent
  • IL-12p70 Detection Antibody
  • IL-12p70 Standard
  • 10X Wash Buffer
  • Buffer D
  • Elution Buffer B

Legal Information

SMC is a registered trademark of Singulex
SMCxPRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Use the power of high sensitivity immunology assays with SMC® technology to precisely characterize low-abundant cytokines like those from Th1/Th2 and Th17 cells.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service