MAT062354318
Matrix Scientific
5-Methyl-4-morpholin-4-ylmethyl-furan-2-carboxylicacid hydrochloride
About This Item
Iupac Name
5-methyl-4-(4-morpholinylmethyl)-2-furoic acid hydrochloride
Inchi Code
1S/C11H15NO4.ClH/c1-8-9(6-10(16-8)11(13)14)7-12-2-4-15-5-3-12;/h6H,2-5,7H2,1H3,(H,13,14);1H
InChI key
FZIMAWVOPGCTNK-UHFFFAOYSA-N
Country of Origin
Product Link
MSDS
https://www.matrixscientific.com/010221.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.