MAN372905805
Manchester Organics
Methyl pentafluoropropionylacetate
About This Item
Iupac Name
methyl 4,4,5,5,5-pentafluoro-3-oxopentanoate
Inchi Code
1S/C6H5F5O3/c1-14-4(13)2-3(12)5(7,8)6(9,10)11/h2H2,1H3
InChI key
FGGMNRZYEBHEIR-UHFFFAOYSA-N
Country of Origin
Product Link
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.