Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAN372905206

Manchester Organics

10-Undecenyltrichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
287.73
MDL number:

Iupac Name

trichloro(10-undecenyl)silane

Inchi Code

1S/C11H21Cl3Si/c1-2-3-4-5-6-7-8-9-10-11-15(12,13)14/h2H,1,3-11H2

InChI key

KFFLNZJAHAUGLE-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Product Link

https://www.manchesterorganics.com/L26103

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library