LABC-1278-960-008-9
Labcon
Refillable Racks for 200-300µL Extended Pipette Tips
96 Rack, 200-300uL, Quantity per Pack: 6, Clear, Teal
Synonym(s):
Rack
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
manufacturer/tradename
Labcon
General description
This is a Labcon branded product.
Per Pack: 6
Per Case: 48
Pack:Case Ratio: 8
Package Type: 96 Rack
Sterile: No
Shelf Life (Months): 72
Shelf Life (Days): 2160
Volume: 200-300uL
Color: Clear, Teal
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.