LABC-1277-960-006-9
Labcon
Refillable Racks for Eclipse 100-1250µL Refill Pipette Tips
96 Rack, 100-1250uL, Quantity per Pack: 6, Clear, Purple, Teal
Synonym(s):
Rack
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
manufacturer/tradename
Labcon
General description
This is a Labcon branded product.
Per Pack: 6
Per Case: 48
Pack:Case Ratio: 8
Package Type: 96 Rack
Sterile: No
Shelf Life (Months): 72
Shelf Life (Days): 2160
Volume: 100-1250uL
Color: Clear, Purple, Teal
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.