Key Documents
LABC-1032-260-000-9
Labcon
Eclipse 100µL Eppendorf-Style Yellow Pipette Tips
Eclipse™ Refill, 100uL, Quantity per Pack: 960, Yellow, Polypropylene
Synonym(s):
Tip, Pipette, Eppendorf
About This Item
PDF Icon
Shipped & Sold By Labcon
A Trusted Marketplace Seller
PDF Icon
Have a question?
Product LABC-1032-260-000-9 is currently not available for purchase. Contact Labcon.
manufacturer/tradename
Labcon
General description
Eclipse™ standard surface pipette tips are made of high quality 100% pure virgin medical grade polypropylene. We pre-test the resin to ensure no extractables are present. Per Pack: 960 Per Case: 9600 Pack:Case Ratio: 10 Package Type: Eclipse™ Refill Sterile: No Shelf Life (Months): 72 Shelf Life (Days): 2160 Volume: 100uL Tip Length (mm): 49.7 Color: Yellow Material: Polypropylene
Legal Information
Eclipse™ is a registered trademark of Labcon.
Choose from one of the most recent versions:
Certificates of Analysis (COA)