KEY465202113
Key Organics / BIONET
2-{[2-(cyclohexylsulfanyl)-5-nitrophenyl]methylidene}propanedinitrile
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2-[2-(cyclohexylsulfanyl)-5-nitrobenzylidene]malononitrile
Inchi Code
1S/C16H15N3O2S/c17-10-12(11-18)8-13-9-14(19(20)21)6-7-16(13)22-15-4-2-1-3-5-15/h6-9,15H,1-5H2
InChI key
OKGKWLQNLVWVQG-UHFFFAOYSA-N
Purity
90%
Country of Origin
GB
Physical Form
Solid
Product Link
https://www.keyorganics.net/sku/JS-0800
Signal Word
Warning
Hazard Codes
MSDS
https://www.keyorganics.net/sds/JS-0800
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.