Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY465198035

Key Organics / BIONET

1-[(4-fluorophenyl)methoxy]-1H-imidazole-2-carbaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
220.2
MDL number:
Product KEY465198035 is currently not available for purchase. Contact Technical Service

Iupac Name

1-[(4-fluorobenzyl)oxy]-1H-imidazole-2-carbaldehyde

Inchi Code

1S/C11H9FN2O2/c12-10-3-1-9(2-4-10)8-16-14-6-5-13-11(14)7-15/h1-7H,8H2

InChI key

LZCYUJRLJGWFGC-UHFFFAOYSA-N

Purity

90%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/6P-020

pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/6P-020


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library