Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY407051039

Key Organics / BIONET

1,1,2,2-tetramethyl-1,2-bis(thiophen-2-yl)disilane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
282.58
MDL number:
Product KEY407051039 is currently not available for purchase. Contact Technical Service

Iupac Name

1,1,2,2-tetramethyl-1,2-di(2-thienyl)disilane

Inchi Code

1S/C12H18S2Si2/c1-15(2,11-7-5-9-13-11)16(3,4)12-8-6-10-14-12/h5-10H,1-4H3

InChI key

FGYSQQKHYAVZEZ-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Low Melting Solid

Product Link

https://www.keyorganics.net/sku/AS-2202

pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/AS-2202


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library