KEY407050960
Key Organics / BIONET
9-benzyl-3-bromo-9H-carbazole
Sign Into View Organizational & Contract Pricing
Select a Size
All Photos(1)
Select a Size
Change View
About This Item
Iupac Name
9-benzyl-3-bromo-9H-carbazole
Inchi Code
1S/C19H14BrN/c20-15-10-11-19-17(12-15)16-8-4-5-9-18(16)21(19)13-14-6-2-1-3-7-14/h1-12H,13H2
InChI key
NMNQVRIUUBFICK-UHFFFAOYSA-N
Purity
95%
Country of Origin
GB
Physical Form
Solid
Product Link
https://www.keyorganics.net/sku/AS-2030
Signal Word
Warning
Hazard Codes
MSDS
https://www.keyorganics.net/sds/AS-2030
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.