KEY158583329
Key Organics / BIONET
4-(4-benzylpiperazin-1-yl)-6-chloro-2-methylpyrimidine
About This Item
mp
94 - 96
Iupac Name
4-(4-benzyl-1-piperazinyl)-6-chloro-2-methylpyrimidine
Inchi Code
1S/C16H19ClN4/c1-13-18-15(17)11-16(19-13)21-9-7-20(8-10-21)12-14-5-3-2-4-6-14/h2-6,11H,7-10,12H2,1H3
InChI key
IOKPDSNVJVSREP-UHFFFAOYSA-N
Purity
Country of Origin
Physical Form
Product Link
Melting Point
Signal Word
Warning
Hazard Codes
Precautionary statements
MSDS
https://www.keyorganics.net/sds/MB-0706
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.