Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY004181634

Key Organics / BIONET

3-(azepan-1-yl)propan-1-amine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
156.27
MDL number:

bp

222C

Iupac Name

3-(1-azepanyl)-1-propanamine

Inchi Code

1S/C9H20N2/c10-6-5-9-11-7-3-1-2-4-8-11/h1-10H2

Inchi Key

FXTRRLIESPAUHT-UHFFFAOYSA-N

storage temp.

Store under inert gas

Purity

95%

Country of Origin

GB

Physical Form

Liquid

Product Link

https://www.keyorganics.net/sku/8P-071

Boiling point

222C

Melting Point


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library