Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY001124005

Key Organics / BIONET

4-[4-(4-fluorophenyl)piperazin-1-yl]aniline

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
271.34
MDL number:
Product KEY001124005 is currently not available for purchase. Contact Technical Service

mp

177 - 179

Iupac Name

4-[4-(4-fluorophenyl)-1-piperazinyl]aniline

Inchi Code

1S/C16H18FN3/c17-13-1-5-15(6-2-13)19-9-11-20(12-10-19)16-7-3-14(18)4-8-16/h1-8H,9-12,18H2

InChI key

FQOITVYFRAXELU-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/10G-527S

Melting Point

177 - 179

pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/10G-527S


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library