Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JWPH95E08DB0

J & W Pharmlab LLC

trans-4-Hydroxy-3-methyl-piperidine-1-carboxylic acid tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
215.29
MDL number:

Iupac Name

tert-butyl (3S,4S)-4-hydroxy-3-methylpiperidine-1-carboxylate

Inchi Code

1S/C11H21NO3/c1-8-7-12(6-5-9(8)13)10(14)15-11(2,3)4/h8-9,13H,5-7H2,1-4H3/t8-,9-/m0/s1

InChI key

PSDMSGJMWVMEMV-IUCAKERBSA-N

Purity

96%

Country of Origin

US

Physical Form

Light Yellow Liquid

Product Link

https://www.jwpharmlab.com/home/product/60R1793

MSDS

https://www.jwpharmlab.com/home/product/60R1793


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library