Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JWPH942F371D

J & W Pharmlab LLC

2-Isopropyl-oxazole-5-boronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
237.11
MDL number:

Iupac Name

2-isopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)oxazole

Inchi Code

1S/C12H20BNO3/c1-8(2)10-14-7-9(15-10)13-16-11(3,4)12(5,6)17-13/h7-8H,1-6H3

InChI key

MGYUOWASNFATFI-UHFFFAOYSA-N

Purity

96%

Country of Origin

US

Physical Form

White Solid

Product Link

https://www.jwpharmlab.com/home/product/32R0292P

MSDS

https://www.jwpharmlab.com/home/product/32R0292P


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library