Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JWPH3249DAE5

J & W Pharmlab LLC

4-(4(5)-Methyl-1H-imidazol-2-yl)-piperidine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
238.16
MDL number:

Iupac Name

4-(4-methyl-1H-imidazol-2-yl)piperidine dihydrochloride

Inchi Code

1S/C9H15N3.2ClH/c1-7-6-11-9(12-7)8-2-4-10-5-3-8;;/h6,8,10H,2-5H2,1H3,(H,11,12);2*1H

InChI key

RFSXQCGSNNQKAQ-UHFFFAOYSA-N

Purity

98%

Country of Origin

US

Physical Form

yellow solid

Product Link

https://www.jwpharmlab.com/home/product/60R0503S

MSDS

https://www.jwpharmlab.com/home/product/60R0503S


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library