Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JWPH3249D7D7

J & W Pharmlab LLC

2-Methyl-4,5,6,7-tetrahydro-oxazolo[4,5-c]pyridine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
174.63
MDL number:

Iupac Name

2-methyl-4,5,6,7-tetrahydro[1,3]oxazolo[4,5-c]pyridine hydrochloride

Inchi Code

1S/C7H10N2O.ClH/c1-5-9-6-4-8-3-2-7(6)10-5;/h8H,2-4H2,1H3;1H

InChI key

SQLHPVHVGOGENV-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Physical Form

White Solid

Product Link

https://www.jwpharmlab.com/home/product/56R0104S

MSDS

https://www.jwpharmlab.com/home/product/56R0104S


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library