JWPH3249CDD5
J & W Pharmlab LLC
2-Trifluoromethyl-azetidine tosylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2-(trifluoromethyl)azetidine 4-methylbenzenesulfonate
Inchi Code
1S/C7H8O3S.C4H6F3N/c1-6-2-4-7(5-3-6)11(8,9)10;5-4(6,7)3-1-2-8-3/h2-5H,1H3,(H,8,9,10);3,8H,1-2H2
InChI key
CDFFBCXHVADOPM-UHFFFAOYSA-N
Purity
96%
Country of Origin
US
Physical Form
Off White Solid
Product Link
https://www.jwpharmlab.com/home/product/25R0106T
MSDS
https://www.jwpharmlab.com/home/product/25R0106T
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.