Key Documents
HS3301NG
Hausser Scientific
Hausser Scientific Counting Chamber - No Grid
Counting chamber, 100 micron cell depth, two counting areas, "V" groove loading, two cover slips
About This Item
PDF Icon
Shipped & Sold By HAUSSER SCIENTIFIC CO.
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Hausser Scientific
General description
Counting chamber, 100 micron cell depth, two counting areas, "V" groove loading, two cover slips
Choose from one of the most recent versions:
Certificates of Analysis (COA)