Key Documents
PCR-384-ABIW
Globe Scientific
Globe Scientific PCR Plates
384-Well PCR Plate, A24 Single Notch design (ABI-Style), White-BOX/10
Select a Size
1 EA
$63.90
$63.90
In StockDetails
Select a Size
Change View
1 EA
$63.90
About This Item
PDF Icon
Shipped & Sold By Globe Scientific LLC
A Trusted Marketplace Seller
PDF Icon
Have a question?
$63.90
In StockDetails
General description
Globe Scientific® PCR Plates are offered in a variety of formats including no skirt, half skirt and full skirt variants in 0.1mL (low profile) and 0.2mL volumes. Available 384 well variants round out this expansive offering. Globe offers a PCR plate compatible with most all commercially available PCR and qPCR instruments.
Features:
  • Ultra-thin wall design (only 0.2mm thick) for optimal temperature transfer
  • Ultra-clear wells for easy sample visualization
  • Compatibility across a broad range of thermal cyclers
  • Easy-to-read alphanumeric indexing
  • Manufactured from high-clarity, ultra-pure polypropylene (meets FDA 21 CFR 177.1520 & USP Class VI)
  • Certified free of detectable DNase, RNase, Human DNA, PCR inhibitors and tested Pyrogen free
  • Raised well rims for optimal sealing with strip caps, sealing mats, or sealing film
  • No skirt, half skirt and full skirt plates available in a variety of formats
  • 0.1mL low profile, standard 0.2mL and 384 well variants
  • Barcoding options available as special order
  • Made in the USA
Choose from one of the most recent versions:
Certificates of Analysis (COA)