Key Documents
LCS-24X13RW
Globe Scientific
Globe Scientific Cryogenic Label Sheets - Labels for Tubes
Label Sheets, Cryo, 24x13mm, for 0.5mL Tubes, Assorted Colors (595 labels in blue, green, violet, red and yellow)-BOX/2975
Select a Size
1 EA
$114.04
$114.04
Available to ship onMarch 22, 2025Details
Select a Size
Change View
1 EA
$114.04
About This Item
PDF Icon
Shipped & Sold By Globe Scientific LLC
A Trusted Marketplace Seller
PDF Icon
Have a question?
$114.04
Available to ship onMarch 22, 2025Details
General description
Cryogenic storage labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196°C (-311°F). Our cryogenic storage labels can be printed or written on and will not bleed, smudge or smear. These labels are offered in a wide variety of sizes and colors on rolls or laser sheets--perfect for distinguishing various specimens after prolonged periods of cryogenic storage.
Features:
  • Able to withstand extreme cold temperatures
  • Can be printed on with a laser printer
  • Can be written on
  • Available on rolls or sheets
  • Full line of colors
  • Various sizes offered
  • Printing instructions and template included
  • Made in the U.S.A.
Choose from one of the most recent versions:
Certificates of Analysis (COA)