Key Documents
7510060BKS
Globe Scientific
Globe Scientific Diamond® Essentials™ Bulk Wide Mouth HDPE Bottles
Bottle, Wide Mouth, Round, HDPE with PP Closure, 60mL, Bulk Packed with Bottles and Caps Bagged Separately, 1000/Case
Select a Size
1 EA
$730.00
$730.00
In StockDetails
Select a Size
Change View
1 EA
$730.00
About This Item
PDF Icon
Shipped & Sold By Globe Scientific LLC
A Trusted Marketplace Seller
PDF Icon
Have a question?
$730.00
In StockDetails
feature
with cap
material
Bottle: high-density polyethylene
General description
Wide mouth bottles facilitate the rapid filling and dispensing of solids, powders, and viscous materials.
Diamond® Essentials™ bulk plastic bottles meet the market need for efficient, effective, and economical packaging options aimed at manual and automated filling operations. Packaged in bulk with caps and bottles bagged separately, no de-capping step is needed before filling Diamond® Essentials™ Bulk Bottles. Available in wide and narrow mouth options with cap and closure sizes that match industry norms, the bottles integrate easily into filling operations.
Diamond® Essentials™ bulk plastic laboratory bottles are precision molded from virgin high density polyethylene (HDPE) resins for consistent wall thickness and smooth surfaces inside and out. These rugged HDPE bottles are resistant to most corrosives and are suitable for freezer storage down to -100ºC. Amber bottle options reduce UV light transmission to protect light-sensitive contents and comply with US Pharmacopeia requirements for light transmission. Polypropylene caps are engineered with an integrated seal ring and semi-buttressed threads for leak-proof closure. These leak-proof bottles are designed and tested to withstand the highest pressure thresholds in the industry.
Choose from one of the most recent versions:
Certificates of Analysis (COA)