Key Documents
4974
Globe Scientific
Globe Scientific Formalin Storage Bags
Bag, Liquid Tight Ziplock for Specimen Storage with Formalin Warning Printing, Saranex, 9" x 12", 250/Pack, 4 Packs/Unit-CS/1000
About This Item
PDF Icon
Shipped & Sold By Globe Scientific LLC
A Trusted Marketplace Seller
PDF Icon
Have a question?
General description
The liquid-tight formalin bag with vapor barrier effectively prevents the transmission of harmful fumes associated with formalin. This unique bag is made by a patented manufacturing process which eliminates leaks. Made from strong 3-mil Serinex, the Bitran Formalin Bag provides long-term protection. For added convenience, the bag is preprinted with a biohazard symbol, formalin warnings, and emergency response block as required by OSHA. The doubletrack zipper provides easy closure and airtight security.
  • Packaged in packs of 250 each, four packs per case.
  • Made from strong 3-mil Serinex
Special Order Item. Lead time is approximately 7 to 14 days.
Choose from one of the most recent versions:
Certificates of Analysis (COA)