Key Documents
110416
Globe Scientific
Globe Scientific Plastic Tubes 12mm x 75mm
Test Tube, 12 x 75mm, (5mL), PS, 1000/Bag, 1 Bag/Box
About This Item
PDF Icon
Shipped & Sold By Globe Scientific LLC
A Trusted Marketplace Seller
PDF Icon
Have a question?
General description
Globe Scientific's plastic culture tubes are produced in polystyrene (PS) or polypropylene (PP). The polystyrene tubes can withstand 1500 RPM during centrifugation. Polypropylene tubes can withstand 3000 RPM and are autoclavable and freezable. Tubes with dual position snap caps (LDPE) provide either aerobic or anaerobic storage of samples. Note - Leave the dual position snap cap in the loose position for aerobic work and in the sealed position for anaerobic cultures.
Choose from one of the most recent versions:
Certificates of Analysis (COA)