100B-TM612
Glas-Col®
Glas-Col® Aluminum Heating Mantle
800 ml
Synonym(s):
heating mantle bottom, heating mantle
Select a Size
$374.00
Available to ship onApril 16, 2025Details
Select a Size
About This Item
$374.00
Available to ship onApril 16, 2025Details
AC/DC
115
feature
CSA approved
heater capacity
350W
manufacturer/tradename
Glas-Col
General description
Aluminum mantle for 800 ml griffin beaker.
- 350 Watts
- 115 Volts
- Max Temperature 400oC
- CSA Approved
- A controller is required for operating mantles
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.