Key Documents
55N-HE01-FLS
Foxx Life Sciences
Foxx Life Sciences EZBio® Single Use Carboy Assembly
EZBio® Single Use Carboy Assembly, 20 L, Dark Amber PP, 120mm Cap,TPE Tubing/Dip Tube, Sterile, 1/EA
About This Item
PDF Icon
Shipped & Sold By Foxx Life Sciences
A Trusted Marketplace Seller
PDF Icon
Have a question?
capacity
20 L
feature
with cap
material
Bottle: polypropylene
manufacturer/tradename
Foxx Life Sciences
General description
EZBio® Carboys' distinctive rectangular shape saves valuable bench space while the molded grips provide a secure hold to facilitate in lifting, handling, and pouring. With easy to read metric graduation marks, certified to ±5% accuracy, these carboys include pre-mounted, high tensile strength, platinum-cured silicone tubing. This assembly provides a flexible two-port filling and venting system, allowing for extractable free, aseptic gas and liquid transfer as required for pharmaceutical, biotechnology, and laboratory fluid transfer applications. These high-quality carboy assemblies are readily able to accept additional fluid transfer components such as aseptic connectors, filters, and manifold integration.
Specifications
  • Autoclavable
  • USP Class VI, FDA Grade materials
  • Clear for high visibility of the product
  • High-impact strength
  • Ergonomic shape
  • Leakproof
  • Gamma Irradiated in the U.S.
Choose from one of the most recent versions:
Certificates of Analysis (COA)