Key Documents
252-1907-OEM
Foxx Life Sciences
Foxx Life Sciences EZwaste® HPLC Solvent Waste System, Secondary Container
HPLC Solvent Waste Safety Tray Secondary Container, for 20L-60L Carboys, 3/PK
About This Item
PDF Icon
Shipped & Sold By Foxx Life Sciences
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Foxx Life Sciences
General description
Foxx Life Sciences' Secondary Container is a perfect solution for preventing leakage and spillage from Foxx Life Sciences 1L, 2.5L, 4L, 5L, and 10L EZwaste® Solvent Waste System containers. Made from HDPE material, these containers are built to be tough and durable. Our Secondary Containers have the ability to hold at least 115% of the container's content.
Check our secondary containers' specifications to see if they fit your container.
Made to Order